Search
Search

OXCT1| Gene Symbol: OXCT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309306.1 | NP_001296235.1 | 4 - 5 | 618 | 62 | |
| XM_023625281.1 | XP_023481049.1 | 620 | 62 | |||
| GenBank mRNA | AB646797.1 | - | 4 - 5 | 618 | 62 | |
Set Membership: |
|