Search
Search

ACTR6| Gene Symbol: ACTR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309319.1 | NP_001296248.1 | 10 - 11 | 1110 | 94 | |
| XM_005606542.3 | XP_005606599.1 | 1087 | 94 | |||
| XM_023631001.1 | XP_023486769.1 | 1280 | 94 | |||
| GenBank mRNA | JL616511.1 | - | 10 - 11 | 1110 | 94 | |
Set Membership: |
|