Search
Search

ELAVL4| Gene Symbol: ELAVL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492528.5 | XP_001492578.1 | 816 | 63 | ||
| XM_005607067.3 | XP_005607124.1 | 1220 | 63 | |||
| XM_014737536.2 | XP_014593022.1 | 1217 | 63 | |||
| XM_023630234.1 | XP_023486002.1 | 638 | 63 | |||
| XM_023630249.1 | XP_023486017.1 | 902 | 63 | |||
| XM_023630259.1 | XP_023486027.1 | 630 | 63 | |||
Set Membership: |
|