Search
Search

TNRC6B| Gene Symbol: TNRC6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736852.2 | XP_014592338.2 | 2772 | 104 | ||
| XM_023631470.1 | XP_023487238.1 | 678 | 104 | |||
| XM_023631471.1 | XP_023487239.1 | 611 | 104 | |||
| XM_023631472.1 | XP_023487240.1 | 508 | 104 | |||
| XM_023631473.1 | XP_023487241.1 | 417 | 104 | |||
| XM_023631474.1 | XP_023487242.1 | 613 | 104 | |||
Set Membership: |
|