Search
Search

STK24| Gene Symbol: STK24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732158.2 | XP_014587644.1 | 414 | 79 | ||
| XM_023621752.1 | XP_023477520.1 | 830 | 79 | |||
| XM_023621753.1 | XP_023477521.1 | 830 | 79 | |||
| XM_023621754.1 | XP_023477522.1 | 830 | 79 | |||
| XM_023621755.1 | XP_023477523.1 | 830 | 79 | |||
| GenBank mRNA | JL627367.1 | - | 2 - 3 | 121 | 79 | |
Set Membership: |
|