Search
Search

GDPD4| Gene Symbol: GDPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645661.1 | XP_023501429.1 | 976 | 95 | ||
| XM_023645662.1 | XP_023501430.1 | 2176 | 95 | |||
| XM_023645663.1 | XP_023501431.1 | 461 | 95 | |||
| XM_023645664.1 | XP_023501432.1 | 669 | 95 | |||
| XM_023645665.1 | XP_023501433.1 | 534 | 95 | |||
| XM_023645666.1 | XP_023501434.1 | 976 | 95 | |||
Set Membership: |
|