Search
Search

TRIM21| Gene Symbol: TRIM21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005612079.3 | XP_005612136.1 | 1289 | 68 | ||
| XM_023645851.1 | XP_023501619.1 | 1109 | 68 | |||
| XM_023645852.1 | XP_023501620.1 | 1313 | 68 | |||
Set Membership: |
|