Search
Search

DCUN1D2| Gene Symbol: DCUN1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023621895.1 | XP_023477663.1 | 764 | 139 | ||
| XM_023621896.1 | XP_023477664.1 | 950 | 139 | |||
| XM_023621897.1 | XP_023477665.1 | 972 | 139 | |||
| XM_023621898.1 | XP_023477666.1 | 2104 | 139 | |||
| XM_023621899.1 | XP_023477667.1 | 2149 | 139 | |||
| XM_023621900.1 | XP_023477668.1 | 1967 | 139 | |||
Set Membership: |
|