Search
Search

UGGT2| Gene Symbol: UGGT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001916487.4 | XP_001916522.1 | 3483 | 100 | ||
| XM_023621730.1 | XP_023477498.1 | 3483 | 100 | |||
| XM_023621731.1 | XP_023477499.1 | 3483 | 100 | |||
| XM_023621732.1 | XP_023477500.1 | 3212 | 100 | |||
| XM_023621734.1 | XP_023477502.1 | 3483 | 100 | |||
Set Membership: |
|