Search
Search

DMXL1| Gene Symbol: DMXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504549.6 | XP_001504599.1 | 619 | 67 | ||
| XM_003362834.4 | XP_003362882.1 | 619 | 67 | |||
| XM_014730724.2 | XP_014586210.1 | 621 | 67 | |||
| XM_014730725.2 | XP_014586211.1 | 621 | 67 | |||
Set Membership: |
|