Search
Search

COMMD10| Gene Symbol: COMMD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503485.6 | XP_001503535.3 | 478 | 74 | ||
| XM_005599502.3 | XP_005599559.1 | 478 | 74 | |||
| XM_023617836.1 | XP_023473604.1 | 478 | 74 | |||