Search
Search

NUDT12| Gene Symbol: NUDT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504597.4 | XP_001504647.1 | 1077 | 76 | ||
| XM_005599516.3 | XP_005599573.1 | 1088 | 76 | |||
| XM_005599517.3 | XP_005599574.1 | 1064 | 76 | |||
Set Membership: |
|