Search
Search

KIAA0825| Gene Symbol: KIAA0825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023617980.1 | XP_023473748.1 | 629 | 107 | ||
| XM_023617982.1 | XP_023473750.1 | 1120 | 107 | |||
| XM_023617983.1 | XP_023473751.1 | 1043 | 107 | |||
| XM_023617984.1 | XP_023473752.1 | 1037 | 107 | |||
| XM_023617985.1 | XP_023473753.1 | 1015 | 107 | |||
| XM_023617986.1 | XP_023473754.1 | 1000 | 107 | |||
| XM_023617987.1 | XP_023473755.1 | 996 | 107 | |||
| XM_023617988.1 | XP_023473756.1 | 917 | 107 | |||
| XM_023617989.1 | XP_023473757.1 | 938 | 107 | |||
| XM_023617990.1 | XP_023473758.1 | 923 | 107 | |||
| XM_023617991.1 | XP_023473759.1 | 1057 | 107 | |||
| XM_023617993.1 | XP_023473761.1 | 980 | 107 | |||
| XM_023617994.1 | XP_023473762.1 | 750 | 107 | |||
| XM_023617995.1 | XP_023473763.1 | 566 | 107 | |||
| XM_023617996.1 | XP_023473764.1 | 860 | 107 | |||
| XM_023617997.1 | XP_023473765.1 | 1150 | 107 | |||
| XM_023617998.1 | XP_023473766.1 | 399 | 107 | |||
| XM_023617999.1 | XP_023473767.1 | 629 | 107 | |||
| XM_023618000.1 | XP_023473768.1 | 629 | 107 | |||
| XM_023618001.1 | XP_023473769.1 | 637 | 107 | |||
| XM_023618002.1 | XP_023473770.1 | 629 | 107 | |||
| XM_023618004.1 | XP_023473772.1 | 629 | 107 | |||
Set Membership: |
|