Search
Search

EDIL3| Gene Symbol: EDIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504634.4 | XP_001504684.2 | 1052 | 91 | ||
| XM_003362850.4 | XP_003362898.2 | 1019 | 91 | |||
| XM_023618037.1 | XP_023473805.1 | 666 | 91 | |||
Set Membership: |
|