Search
Search

RUFY4| Gene Symbol: RUFY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023642306.1 | XP_023498074.1 | 2818 | 88 | ||
| XM_023642307.1 | XP_023498075.1 | 3791 | 88 | |||
| XM_023642308.1 | XP_023498076.1 | 2654 | 88 | |||
| XM_023642309.1 | XP_023498077.1 | 2327 | 88 | |||
| XM_023642310.1 | XP_023498078.1 | 2254 | 88 | |||
| XM_023642312.1 | XP_023498080.1 | 3039 | 88 | |||
Set Membership: |
|