Search
Search

CPED1| Gene Symbol: CPED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609330.3 | XP_005609387.1 | 2319 | 61 | ||
| XM_014739102.2 | XP_014594588.1 | 2680 | 61 | |||
| XM_023639557.1 | XP_023495325.1 | 2223 | 61 | |||
| XM_023639558.1 | XP_023495326.1 | 3375 | 61 | |||
Set Membership: |
|