Search
Search

ZNF800| Gene Symbol: ZNF800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023639612.1 | XP_023495380.1 | 1072 | 74 | ||
| XM_023639614.1 | XP_023495382.1 | 450 | 74 | |||
| XM_023639615.1 | XP_023495383.1 | 983 | 74 | |||
| XM_023639616.1 | XP_023495384.1 | 1069 | 74 | |||
| XM_023639617.1 | XP_023495385.1 | 1066 | 74 | |||
Set Membership: |
|