Search
Search

AHCYL2| Gene Symbol: AHCYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001917111.5 | XP_001917146.2 | 1830 | 123 | ||
| XM_005609359.3 | XP_005609416.2 | 2048 | 123 | |||
| XM_005609360.3 | XP_005609417.2 | 2044 | 123 | |||
| XM_014739070.2 | XP_014594556.2 | 1978 | 123 | |||
| XM_014739072.2 | XP_014594558.1 | 2118 | 123 | |||
| XM_014739073.2 | XP_014594559.1 | 2115 | 123 | |||
| XM_023639663.1 | XP_023495431.1 | 1893 | 123 | |||
| XM_023639664.1 | XP_023495432.1 | 1889 | 123 | |||
| XM_023639665.1 | XP_023495433.1 | 1980 | 123 | |||
| XM_023639666.1 | XP_023495434.1 | 1975 | 123 | |||
| XM_023639667.1 | XP_023495435.1 | 1645 | 123 | |||
Set Membership: |
|