Search
Search
| Gene Symbol: BHMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504652.4 | XP_001504702.1 | 1174 | 60 | ||
| XM_023618111.1 | XP_023473879.1 | 1442 | 60 | |||
| XM_023618112.1 | XP_023473880.1 | 1027 | 60 | |||
Set Membership: |
|