Search
Search

CFAP36| Gene Symbol: CFAP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496377.6 | XP_001496427.1 | 349 | 160 | ||
| XM_005599974.3 | XP_005600031.1 | 349 | 160 | |||
| XM_023619070.1 | XP_023474838.1 | 352 | 160 | |||
| XM_023619071.1 | XP_023474839.1 | 352 | 160 | |||
| GenBank mRNA | JL640506.1 | - | 2 - 3 | 178 | 160 | |
Set Membership: |
|