Search
Search

FOXN2| Gene Symbol: FOXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023619116.1 | XP_023474884.1 | 919 | 98 | ||
| XM_023619117.1 | XP_023474885.1 | 850 | 98 | |||
| XM_023619118.1 | XP_023474886.1 | 928 | 98 | |||
| XM_023619119.1 | XP_023474887.1 | 817 | 98 | |||
| XM_023619120.1 | XP_023474888.1 | 1111 | 98 | |||