Search
Search

RMDN2| Gene Symbol: RMDN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600050.3 | XP_005600107.1 | 612 | 86 | ||
| XM_023619221.1 | XP_023474989.1 | 580 | 86 | |||
| XM_023619223.1 | XP_023474991.1 | 695 | 86 | |||
| GenBank mRNA | JL640792.1 | - | 2 - 3 | 280 | 86 | |
Set Membership: |
|