Search
Search

DPYSL5| Gene Symbol: DPYSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362905.4 | XP_003362953.1 | 1164 | 99 | ||
| XM_014731254.2 | XP_014586740.1 | 1247 | 99 | |||
| XM_023619357.1 | XP_023475125.1 | 808 | 99 | |||
Set Membership: |
|