Search
Search

ASXL2| Gene Symbol: ASXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001918196.4 | XP_001918231.2 | 522 | 114 | ||
| XM_005600159.3 | XP_005600216.2 | 519 | 114 | |||
| XM_023619373.1 | XP_023475141.1 | 339 | 114 | |||
| XM_023619374.1 | XP_023475142.1 | 249 | 114 | |||
Set Membership: |
|