Search
Search

WDR35| Gene Symbol: WDR35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503372.6 | XP_001503422.2 | 3100 | 67 | ||
| XM_023619470.1 | XP_023475238.1 | 3134 | 67 | |||
| XM_023619471.1 | XP_023475239.1 | 2932 | 67 | |||
| XM_023619472.1 | XP_023475240.1 | 2967 | 67 | |||
Set Membership: |
|