Search
Search

C16H3orf20| Gene Symbol: C16H3orf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014731393.2 | XP_014586879.2 | 2795 | 114 | ||
| XM_023619914.1 | XP_023475682.1 | 2164 | 114 | |||
Set Membership: |
|