Search
Search

BRPF1| Gene Symbol: BRPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014731425.2 | XP_014586911.1 | 2220 | 63 | ||
| XM_014731426.2 | XP_014586912.1 | 2219 | 63 | |||
| XM_014731427.1 | XP_014586913.1 | 1856 | 63 | |||
| XM_014731428.1 | XP_014586914.1 | 1856 | 63 | |||
| XM_014731429.1 | XP_014586915.1 | 1856 | 63 | |||
| XM_014731430.1 | XP_014586916.1 | 1856 | 63 | |||
| XM_014731431.1 | XP_014586917.1 | 1856 | 63 | |||
| XM_014731432.1 | XP_014586918.1 | 1856 | 63 | |||
| XM_014731433.2 | XP_014586919.1 | 2222 | 63 | |||
| XM_023620016.1 | XP_023475784.1 | 1856 | 63 | |||
Set Membership: |
|