Search
Search

FRMD4B| Gene Symbol: FRMD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001916183.4 | XP_001916218.1 | 719 | 85 | ||
| XM_023620090.1 | XP_023475858.1 | 455 | 85 | |||
| XM_023620091.1 | XP_023475859.1 | 638 | 85 | |||
| XM_023620092.1 | XP_023475860.1 | 460 | 85 | |||
| XM_023620093.1 | XP_023475861.1 | 796 | 85 | |||