Search
Search

ITIH1| Gene Symbol: ITIH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492526.4 | XP_001492576.1 | 1971 | 91 | ||
| XM_014731620.2 | XP_014587106.1 | 1971 | 91 | |||
| XM_014731621.2 | XP_014587107.1 | 1945 | 91 | |||
| XM_014731622.2 | XP_014587108.1 | 1945 | 91 | |||
Set Membership: |
|