Search
Search

DNAH1| Gene Symbol: DNAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001915893.5 | XP_001915928.3 | 9142 | 55 | ||
| XM_023620319.1 | XP_023476087.1 | 9141 | 55 | |||
| XM_023620321.1 | XP_023476089.1 | 8369 | 55 | |||
| XM_023620322.1 | XP_023476090.1 | 7409 | 55 | |||
| XM_023620323.1 | XP_023476091.1 | 9142 | 55 | |||
Set Membership: |
|