Search
Search

TCAIM| Gene Symbol: TCAIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501325.6 | XP_001501375.1 | 471 | 67 | ||
| XM_005600778.3 | XP_005600835.1 | 518 | 67 | |||
| XM_005600779.3 | XP_005600836.1 | 501 | 67 | |||
| XM_005600780.3 | XP_005600837.1 | 550 | 67 | |||
| XM_014731727.2 | XP_014587213.2 | 519 | 67 | |||
| XM_023620623.1 | XP_023476391.1 | 504 | 67 | |||
| GenBank mRNA | JL623474.1 | - | 1 - 2 | 56 | 67 | |
Set Membership: |
|