Search
Search

ACVR2B| Gene Symbol: ACVR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023620724.1 | XP_023476492.1 | 503 | 92 | ||
| XM_023620725.1 | XP_023476493.1 | 2545 | 92 | |||
| XM_023620726.1 | XP_023476494.1 | 2546 | 92 | |||
| XM_023620727.1 | XP_023476495.1 | 500 | 92 | |||
Set Membership: |
|