Search
Search
| Gene Symbol: THRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600897.3 | XP_005600954.1 | 748 | 95 | ||
| XM_005600898.3 | XP_005600955.1 | 684 | 95 | |||
| XM_023620842.1 | XP_023476610.1 | 1004 | 95 | |||
| XM_023620843.1 | XP_023476611.1 | 878 | 95 | |||
| XM_023620844.1 | XP_023476612.1 | 761 | 95 | |||
| XM_023620845.1 | XP_023476613.1 | 3359 | 95 | |||
| XM_023620846.1 | XP_023476614.1 | 665 | 95 | |||
| XM_023620847.1 | XP_023476615.1 | 913 | 95 | |||
| XM_023620848.1 | XP_023476616.1 | 1109 | 95 | |||
| XM_023620849.1 | XP_023476617.1 | 1011 | 95 | |||
| XM_023620850.1 | XP_023476618.1 | 487 | 95 | |||
Set Membership: |
|