Search
Search

ARMC8| Gene Symbol: ARMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003363167.4 | XP_003363215.2 | 2645 | 88 | ||
| XM_003363168.4 | XP_003363216.2 | 2721 | 88 | |||
| XM_005600950.3 | XP_005601007.1 | 2628 | 88 | |||
| XM_005600951.3 | XP_005601008.1 | 1906 | 88 | |||
| XM_005600952.3 | XP_005601009.1 | 1856 | 88 | |||
| XM_023620998.1 | XP_023476766.1 | 2556 | 88 | |||