Search
Search

TM4SF18| Gene Symbol: TM4SF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600977.3 | XP_005601034.1 | 582 | 85 | ||
| XM_005600978.3 | XP_005601035.1 | 540 | 85 | |||
| XM_014731911.2 | XP_014587397.1 | 1089 | 85 | |||
| XM_014731912.2 | XP_014587398.1 | 583 | 85 | |||
Set Membership: |
|