Search
Search

IFT88| Gene Symbol: IFT88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005601085.3 | XP_005601142.1 | 1988 | 85 | ||
| XM_014731953.2 | XP_014587439.1 | 1988 | 85 | |||
| GenBank mRNA | JL632636.1 | - | 4 - 5 | 322 | 85 | |
Set Membership: |
|