Search
Search

COL6A3| Gene Symbol: COL6A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014740385.2 | XP_014595871.2 | 6314 | 104 | ||
| XM_023642645.1 | XP_023498413.1 | 6937 | 104 | |||
| XM_023642646.1 | XP_023498414.1 | 6332 | 104 | |||
| XM_023642647.1 | XP_023498415.1 | 5711 | 104 | |||
| XM_023642648.1 | XP_023498416.1 | 5111 | 104 | |||
Set Membership: |
|