Search
Search
| Gene Symbol: SCLY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023642695.1 | XP_023498463.1 | 326 | 106 | ||
| XM_023642696.1 | XP_023498464.1 | 325 | 106 | |||
| XM_023642697.1 | XP_023498465.1 | 325 | 106 | |||
| XM_023642698.1 | XP_023498466.1 | 323 | 106 | |||
| XM_023642699.1 | XP_023498467.1 | 327 | 106 | |||
Set Membership: |
|