Search
Search
| Gene Symbol: BID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610818.3 | XP_005610875.1 | 803 | 104 | ||
| XM_023642857.1 | XP_023498625.1 | 845 | 104 | |||
| XM_023642858.1 | XP_023498626.1 | 781 | 104 | |||
Set Membership: |
|