Search
Search

C6H12orf4| Gene Symbol: C6H12orf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643004.1 | XP_023498772.1 | 1674 | 68 | ||
| XM_023643005.1 | XP_023498773.1 | 1852 | 68 | |||
| XM_023643006.1 | XP_023498774.1 | 1960 | 68 | |||
| XM_023643007.1 | XP_023498775.1 | 1729 | 68 | |||
| GenBank mRNA | HQ890302.1 | - | 7 - 8 | 828 | 68 | |
Set Membership: |
|