Search
Search

PIBF1| Gene Symbol: PIBF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023621681.1 | XP_023477449.1 | 491 | 96 | ||
| XM_023621682.1 | XP_023477450.1 | 486 | 96 | |||
| XM_023621683.1 | XP_023477451.1 | 496 | 96 | |||
Set Membership: |
|