Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ec06957899_g1
See other NEB GE Assays ›
Gene Symbol:
NEB
Gene Name:
nebulin
Gene Aliases:
-
Chromosome Location:
Chr.18: 33649007 - 33862519 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID
Ec06957899_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: NEB
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_023622873.1
XP_023478641.1
12524
79
XM_023622874.1
XP_023478642.1
12524
79
XM_023622875.1
XP_023478643.1
12524
79
XM_023622876.1
XP_023478644.1
12524
79
XM_023622877.1
XP_023478645.1
12524
79
XM_023622878.1
XP_023478646.1
12524
79
XM_023622879.1
XP_023478647.1
12524
79
XM_023622880.1
XP_023478648.1
12524
79
XM_023622881.1
XP_023478649.1
12524
79
XM_023622882.1
XP_023478650.1
12524
79
XM_023622883.1
XP_023478651.1
12524
79
XM_023622884.1
XP_023478652.1
12524
79
XM_023622885.1
XP_023478653.1
12524
79
XM_023622886.1
XP_023478654.1
12524
79
XM_023622887.1
XP_023478655.1
12524
79
XM_023622888.1
XP_023478656.1
12524
79
XM_023622889.1
XP_023478657.1
12524
79
XM_023622890.1
XP_023478658.1
12524
79
XM_023622891.1
XP_023478659.1
12524
79
XM_023622892.1
XP_023478660.1
12524
79
XM_023622893.1
XP_023478661.1
12524
79
XM_023622894.1
XP_023478662.1
12524
79
XM_023622895.1
XP_023478663.1
12524
79
XM_023622896.1
XP_023478664.1
12524
79
XM_023622897.1
XP_023478665.1
12524
79
XM_023622898.1
XP_023478666.1
12524
79
XM_023622899.1
XP_023478667.1
12524
79
XM_023622900.1
XP_023478668.1
12524
79
XM_023622901.1
XP_023478669.1
12524
79
XM_023622902.1
XP_023478670.1
12524
79
XM_023622903.1
XP_023478671.1
12524
79
XM_023622904.1
XP_023478672.1
12524
79
XM_023622905.1
XP_023478673.1
11066
79
XM_023622906.1
XP_023478674.1
12524
79
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Equus caballus
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline