Search
Search

TANC1| Gene Symbol: TANC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732481.2 | XP_014587967.1 | 2993 | 115 | ||
| XM_023622965.1 | XP_023478733.1 | 3657 | 115 | |||
| XM_023622966.1 | XP_023478734.1 | 3658 | 115 | |||
| XM_023622967.1 | XP_023478735.1 | 3638 | 115 | |||
| XM_023622968.1 | XP_023478736.1 | 3639 | 115 | |||
| XM_023622969.1 | XP_023478737.1 | 5267 | 115 | |||
| XM_023622970.1 | XP_023478738.1 | 3383 | 115 | |||
| XM_023622971.1 | XP_023478739.1 | 3556 | 115 | |||
| XM_023622972.1 | XP_023478740.1 | 3396 | 115 | |||
| XM_023622973.1 | XP_023478741.1 | 3377 | 115 | |||
| XM_023622974.1 | XP_023478742.1 | 3708 | 115 | |||
| XM_023622975.1 | XP_023478743.1 | 3302 | 115 | |||
| XM_023622976.1 | XP_023478744.1 | 3603 | 115 | |||
| XM_023622977.1 | XP_023478745.1 | 3016 | 115 | |||
| XM_023622978.1 | XP_023478746.1 | 3805 | 115 | |||
| XM_023622979.1 | XP_023478747.1 | 2923 | 115 | |||
| XM_023622980.1 | XP_023478748.1 | 2884 | 115 | |||
Set Membership: |
|