Search
Search

GPR155| Gene Symbol: GPR155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001917170.4 | XP_001917205.2 | 2540 | 82 | ||
| XM_005601590.2 | XP_005601647.2 | 2742 | 82 | |||
| XM_014732539.2 | XP_014588025.2 | 2658 | 82 | |||
| XM_023622025.1 | XP_023477793.1 | 2599 | 82 | |||
Set Membership: |
|