Search
Search

FASTKD2| Gene Symbol: FASTKD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023622493.1 | XP_023478261.1 | 1908 | 83 | ||
| XM_023622494.1 | XP_023478262.1 | 2099 | 83 | |||
| XM_023622495.1 | XP_023478263.1 | 1908 | 83 | |||