Search
Search

EPHB3| Gene Symbol: EPHB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023623445.1 | XP_023479213.1 | 2238 | 72 | ||
| XM_023623446.1 | XP_023479214.1 | 2229 | 72 | |||
| XM_023623447.1 | XP_023479215.1 | 2238 | 72 | |||
| XM_023623448.1 | XP_023479216.1 | 2229 | 72 | |||
Set Membership: |
|