Search
Search

ACAP2| Gene Symbol: ACAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005602103.3 | XP_005602160.3 | 2294 | 66 | ||
| XM_023623531.1 | XP_023479299.1 | 2468 | 66 | |||
| XM_023623532.1 | XP_023479300.1 | 2455 | 66 | |||
| XM_023623533.1 | XP_023479301.1 | 2446 | 66 | |||
| XM_023623534.1 | XP_023479302.1 | 2433 | 66 | |||
| XM_023623535.1 | XP_023479303.1 | 2415 | 66 | |||
| XM_023623536.1 | XP_023479304.1 | 2360 | 66 | |||
| XM_023623537.1 | XP_023479305.1 | 2278 | 66 | |||
| XM_023623538.1 | XP_023479306.1 | 2269 | 66 | |||
Set Membership: |
|