Search
Search

CYB5RL| Gene Symbol: CYB5RL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492765.5 | XP_001492815.2 | 971 | 73 | ||
| XM_005607026.3 | XP_005607083.1 | 4052 | 73 | |||
| XM_005607027.3 | XP_005607084.1 | 973 | 73 | |||
| XM_005607028.3 | XP_005607085.1 | 975 | 73 | |||
| XM_005607029.3 | XP_005607086.1 | 971 | 73 | |||
| XM_014737462.2 | XP_014592948.1 | 982 | 73 | |||
| XM_014737463.2 | XP_014592949.1 | 973 | 73 | |||
| XM_023629381.1 | XP_023485149.1 | 1239 | 73 | |||
| XM_023629391.1 | XP_023485159.1 | 3863 | 73 | |||
| XM_023629397.1 | XP_023485165.1 | 583 | 73 | |||
| XM_023629406.1 | XP_023485174.1 | 972 | 73 | |||
| XM_023629411.1 | XP_023485179.1 | 450 | 73 | |||
| XM_023629454.1 | XP_023485222.1 | 973 | 73 | |||
| XM_023629477.1 | XP_023485245.1 | 975 | 73 | |||
Set Membership: |
|