Search
Search

CCDC17| Gene Symbol: CCDC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023630976.1 | XP_023486744.1 | 1669 | 102 | ||
| XM_023630990.1 | XP_023486758.1 | 1774 | 102 | |||
| XM_023630998.1 | XP_023486766.1 | 1115 | 102 | |||
Set Membership: |
|