Search
Search

FOXJ3| Gene Symbol: FOXJ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607147.3 | XP_005607204.1 | 1874 | 66 | ||
| XM_023632263.1 | XP_023488031.1 | 1849 | 66 | |||
| XM_023632267.1 | XP_023488035.1 | 1899 | 66 | |||
| XM_023632279.1 | XP_023488047.1 | 1747 | 66 | |||
Set Membership: |
|